ContactUsLinkComponent
TLC Plastic Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, PEI-cellulose, matrix, binder, not applicable, fluoresc ent indicator 1 * 25 items
Catalog # SIALZ740237-25EA
Supplier:  Merck
TLC Plastic Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, PEI-cellulose, matrix, binder, not applicable, fluoresc ent indicator 1 * 25 items
Catalog # SIALZ740237-25EA
Supplier:  Merck
Frequently Bought Together