ContactUsLinkComponent
HPTLC (High Performance TLC) Aluminum Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix (C18), b inder, Polymeric, fluorescent indicator 1 * 20 items
Catalog # SIALZ740234-20EA
Supplier:  Merck
HPTLC (High Performance TLC) Aluminum Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix (C18), b inder, Polymeric, fluorescent indicator 1 * 20 items
Catalog # SIALZ740234-20EA
Supplier:  Merck
Frequently Bought Together