ContactUsLinkComponent
HPTLC (High Performance TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 20 cm, silica gel 60 matrix, binder, Po lymeric, fluorescent indicator 1 * 50 items
Catalog # SIALZ740220-50EA
Supplier:  Merck
HPTLC (High Performance TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 20 cm, silica gel 60 matrix, binder, Po lymeric, fluorescent indicator 1 * 50 items
Catalog # SIALZ740220-50EA
Supplier:  Merck
Frequently Bought Together