ContactUsLinkComponent
PTLC (Preparative TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeri c, fluorescent indicator 1 * 15 items
Catalog # SIALZ740216-15EA
Supplier:  Merck
PTLC (Preparative TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeri c, fluorescent indicator 1 * 15 items
Catalog # SIALZ740216-15EA
Supplier:  Merck
Frequently Bought Together