Lower part Reference 2 single-channel 100µl yellow variable: 10-100µl, fixed: 25µl/50µl/100µl 1 * 1 items
Catalog # EPPE4920605.007
Supplier: EPPENDORF
Lower part Reference 2 single-channel 100µl yellow variable: 10-100µl, fixed: 25µl/50µl/100µl 1 * 1 items
Catalog # EPPE4920605.007
Supplier: EPPENDORF
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
VWR® Traceable® Digital Thermo-Hygrometer
Supplier: VWR Collection
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR® MiniStar/MiniStar blueline, Microcentrifuge
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Eppendorf® Pipette Holder System
Supplier: EPPENDORF
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
3-Packs, Research® plus (General Lab Product)
Supplier: EPPENDORF
Accessories for water purification systems, arium®
Supplier: Sartorius
Pipette stand, linear
Supplier: Sartorius
Funnels with short stem, DURAN®
Supplier: DWK Life Sciences
Accessories for marco
Supplier: Brand
VWR® Facial Tissues
Supplier: VWR Collection