ContactUsLinkComponent
Connectors for Big-Star/Maxi-Desiccators, For safe and fix connection of stacked SICCO Desiccators without boring. Consisting of fou r pairs of braces, each pair with one screw and nut, four silicone o-rings, one hexagon wrench, operating instruction 1 * 1 items
Catalog # BOHLV1954-07
Supplier:  Bohlender
undefined
Connectors for Big-Star/Maxi-Desiccators, For safe and fix connection of stacked SICCO Desiccators without boring. Consisting of fou r pairs of braces, each pair with one screw and nut, four silicone o-rings, one hexagon wrench, operating instruction 1 * 1 items
Catalog # BOHLV1954-07
Supplier:  Bohlender
Frequently Bought Together