ContactUsLinkComponent
Big-Star-Desiccator Protect, Aluminium frame, orange panels made of acrylic glass reduce light incidence and protect against ultravi olet radiation, including two shelves made of stainless steel, tray and hygrometer, usable with up to 8 shelves. 1 * 1 items
Catalog # BOHLV1926-07
Supplier:  Bohlender
undefined
Big-Star-Desiccator Protect, Aluminium frame, orange panels made of acrylic glass reduce light incidence and protect against ultravi olet radiation, including two shelves made of stainless steel, tray and hygrometer, usable with up to 8 shelves. 1 * 1 items
Catalog # BOHLV1926-07
Supplier:  Bohlender
Frequently Bought Together