ContactUsLinkComponent
Threaded Couplings T, Freely turnable screw caps with glthreads made of PPS and PTFE/FKM-sealing. T-Fitting made of PTFE. For connec ting two glthreads of the same size. The product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLH908-14
Supplier:  Bohlender
undefined
Threaded Couplings T, Freely turnable screw caps with glthreads made of PPS and PTFE/FKM-sealing. T-Fitting made of PTFE. For connec ting two glthreads of the same size. The product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLH908-14
Supplier:  Bohlender
Frequently Bought Together