ContactUsLinkComponent
Tri-Clamp Gaskets, Silicone ring with double-sided, elastic washer made of PTFE. Suitable for Tri-Clamp connections as per DIN 32 67 6. Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD930-04
Supplier:  Bohlender
undefined
Tri-Clamp Gaskets, Silicone ring with double-sided, elastic washer made of PTFE. Suitable for Tri-Clamp connections as per DIN 32 67 6. Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD930-04
Supplier:  Bohlender
Frequently Bought Together