ContactUsLinkComponent
Tri-Clamp GL-Stopcocks, 2-Way stopcock made of PTFE with straight bore, one Tri-Clamp neck as per DIN 32 676 and one GL-threaded nec k. Cylindrical stopcock made of PTFE for reliable tightness and grip made of PP for marking the flow direction. Unive 1 * 1 items
Catalog # BOHLD917-24
Supplier:  Bohlender
undefined
Tri-Clamp GL-Stopcocks, 2-Way stopcock made of PTFE with straight bore, one Tri-Clamp neck as per DIN 32 676 and one GL-threaded nec k. Cylindrical stopcock made of PTFE for reliable tightness and grip made of PP for marking the flow direction. Unive 1 * 1 items
Catalog # BOHLD917-24
Supplier:  Bohlender
Frequently Bought Together