ContactUsLinkComponent
Multiple Distributor Tri-Clamp, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to two or three GL-threaded necks . Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD911-22
Supplier:  Bohlender
undefined
Multiple Distributor Tri-Clamp, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to two or three GL-threaded necks . Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD911-22
Supplier:  Bohlender
Frequently Bought Together