ContactUsLinkComponent
GL-Reductions, Red screw cap made of PBTP, movable reduction body made of PP with O-ring made of FKM for transition to glthreads. Th e body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fi 1 * 1 items
Catalog # BOHLD782-16
Supplier:  Bohlender
undefined
GL-Reductions, Red screw cap made of PBTP, movable reduction body made of PP with O-ring made of FKM for transition to glthreads. Th e body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fi 1 * 1 items
Catalog # BOHLD782-16
Supplier:  Bohlender
Frequently Bought Together