ContactUsLinkComponent
Distributors for Canisters EX, Screw cap and movable body with GL-threaded necks made of conductive PTFE-EX. Without earthing conne ction. Earthing is made via a conductive canister. Connection of tubing made by BOLA Laboratory Screw Joints EX. 1 * 1 items
Catalog # BOHLD772-32
Supplier:  Bohlender
undefined
Distributors for Canisters EX, Screw cap and movable body with GL-threaded necks made of conductive PTFE-EX. Without earthing conne ction. Earthing is made via a conductive canister. Connection of tubing made by BOLA Laboratory Screw Joints EX. 1 * 1 items
Catalog # BOHLD772-32
Supplier:  Bohlender
Frequently Bought Together