ContactUsLinkComponent
Guide Sleeve, Suitable for BOLA Special Stirrer Bearings with ground joint ns29/32 for shaft O. D. 8 or 10 mm, Cat. No. C 430-20 and C 430-28 1 * 1 items
Catalog # BOHLC960-01
Supplier:  Bohlender
undefined
Guide Sleeve, Suitable for BOLA Special Stirrer Bearings with ground joint ns29/32 for shaft O. D. 8 or 10 mm, Cat. No. C 430-20 and C 430-28 1 * 1 items
Catalog # BOHLC960-01
Supplier:  Bohlender
Frequently Bought Together