ContactUsLinkComponent
Receiver P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 for shaft O. D. 8 mm, Cat. No. C 520-24 as well as BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 10 mm, Cat. No. C 520-2 1 * 1 items
Catalog # BOHLC936-02
Supplier:  Bohlender
undefined
Receiver P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 for shaft O. D. 8 mm, Cat. No. C 520-24 as well as BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 10 mm, Cat. No. C 520-2 1 * 1 items
Catalog # BOHLC936-02
Supplier:  Bohlender
Frequently Bought Together