ContactUsLinkComponent
Magnetic Stirrer Heads (P-MRK) with Flat Flange, Ideal stirrer head for PTFE-jacketed stirrer shafts from BOLA. Consisting of capsul ed drive shaft (stainless steel) with ball bearings, rotor and lower bearing made of PEEK-Compound and a Hastelloy® r 1 * 1 items
Katalogové číslo BOHLC532-22
Supplier:  Bohlender
undefined
Magnetic Stirrer Heads (P-MRK) with Flat Flange, Ideal stirrer head for PTFE-jacketed stirrer shafts from BOLA. Consisting of capsul ed drive shaft (stainless steel) with ball bearings, rotor and lower bearing made of PEEK-Compound and a Hastelloy® r 1 * 1 items
Katalogové číslo BOHLC532-22
Supplier:  Bohlender
Frequently Bought Together