ContactUsLinkComponent
Mini-Propeller Stirrer Shafts, PTFE-jacketed stainless steel shaft, propeller completely made of PTFE with three 45° angled angular blades. Universal chemical resistance since the product is only exposed to PTFE. 1 * 1 items
Katalogové číslo BOHLC482-08
Supplier:  Bohlender
undefined
Mini-Propeller Stirrer Shafts, PTFE-jacketed stainless steel shaft, propeller completely made of PTFE with three 45° angled angular blades. Universal chemical resistance since the product is only exposed to PTFE. 1 * 1 items
Katalogové číslo BOHLC482-08
Supplier:  Bohlender
Frequently Bought Together