ContactUsLinkComponent
Flat Flange Reaction Vessels, Made of PTFE thick wall with round bottom and smooth interior surface, flat flange with circular groo ve as per DIN 12 214. Can be heated by a thermostat or an electrical heating mantle. 1 * 1 items
Catalog # BOHLB271-06
Supplier:  Bohlender
undefined
Flat Flange Reaction Vessels, Made of PTFE thick wall with round bottom and smooth interior surface, flat flange with circular groo ve as per DIN 12 214. Can be heated by a thermostat or an electrical heating mantle. 1 * 1 items
Catalog # BOHLB271-06
Supplier:  Bohlender
Frequently Bought Together