ContactUsLinkComponent
Screw In Connector with Ground Joint EX, Made of static dissipative PTFE-EX. For connection to BOLA Reactor Lids. Connection with gr ound socket. With hexagonal gripping surface in standard wrench size. Earthing is made via a connection on the reacto 1 * 1 items
Catalog # BOHLB174-06
Supplier:  Bohlender
undefined
Screw In Connector with Ground Joint EX, Made of static dissipative PTFE-EX. For connection to BOLA Reactor Lids. Connection with gr ound socket. With hexagonal gripping surface in standard wrench size. Earthing is made via a connection on the reacto 1 * 1 items
Catalog # BOHLB174-06
Supplier:  Bohlender
Frequently Bought Together