ContactUsLinkComponent
EC HPLC column EC 75/4 NUCLEOSIL 100-5 C18 HD length: 75 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720973.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 75/4 NUCLEOSIL 100-5 C18 HD length: 75 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720973.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together