ContactUsLinkComponent
Safety laced boot, ALESSIO Steel Mid ESD S3, size EU47 / UK12 1 * 1 PAIR
Catalog # ELTN767641/47
Supplier:  ELTEN
undefined
Safety laced boot, ALESSIO Steel Mid ESD S3, size EU47 / UK12 1 * 1 PAIR
Catalog # ELTN767641/47
Supplier:  ELTEN