ContactUsLinkComponent
Precision thermometer, similar DIN, 1 257,51 53,50 119,74 -5/0+100:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, Richter-top finish with button, 420x8-9mm, calibrated, with DKD-Certificate (DAkkS 1 * 1 items
Catalog # AMARL25948CBDKD4CL
Supplier:  Amarell
Precision thermometer, similar DIN, 1 257,51 53,50 119,74 -5/0+100:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, Richter-top finish with button, 420x8-9mm, calibrated, with DKD-Certificate (DAkkS 1 * 1 items
Catalog # AMARL25948CBDKD4CL
Supplier:  Amarell
Frequently Bought Together