ContactUsLinkComponent
The nitric oxide detection kit is a colorimetric assay for the indirect determination of nitric oxide concentrations based on the enzymatic conversion of nitrate to nitrite by a nitrate reductase. 1 * 1 KIT
Catalog # OZBINOS0500
Supplier:  OZ BIOSCIENCES
The nitric oxide detection kit is a colorimetric assay for the indirect determination of nitric oxide concentrations based on the enzymatic conversion of nitrate to nitrite by a nitrate reductase. 1 * 1 KIT
Catalog # OZBINOS0500
Supplier:  OZ BIOSCIENCES
Frequently Bought Together