ContactUsLinkComponent
Rabbit monoclonal antibody raised against synthetic peptide of human SMC1A. 1 * 100 µG
Catalog # ABNOMAB20722
Supplier:  Abnova
Rabbit monoclonal antibody raised against synthetic peptide of human SMC1A. 1 * 100 µG
Catalog # ABNOMAB20722
Supplier:  Abnova
Frequently Bought Together