ContactUsLinkComponent
Flanged Corrugated Tubing, Corrugated tubing with circular corrugations around the longitudinal axis and flanged end pieces made of PFA, with two preassembled screw caps with GL thread made of PBTP and washers made of silicone for direct connection t 1 * 1 items
Catalog # BOHLS1880-10
Supplier:  Bohlender
undefined
Flanged Corrugated Tubing, Corrugated tubing with circular corrugations around the longitudinal axis and flanged end pieces made of PFA, with two preassembled screw caps with GL thread made of PBTP and washers made of silicone for direct connection t 1 * 1 items
Catalog # BOHLS1880-10
Supplier:  Bohlender
Frequently Bought Together