Antistatic Explosion-Proof Tubing, Very good electric conductivity due to a special “antistatic compound” made of pure PTFE and fine st, highly pure carbon dust (less than 2,5%). Colour: black. 1 * 1 items
Catalog # BOHLS1827-34
Supplier: Bohlender
Frequently Bought Together