ContactUsLinkComponent
Flat Flange Lids, Suitable for reaction vessels with SCHOTT flat flange, with center neck socket NS 29/32, lateral necks with ground joint sockets or GL-thread. Bottom side with centring collar for non-slipping alignment on the vessel. All lateral c 1 * 1 items
Catalog # BOHLB210-22
Supplier:  Bohlender
undefined
Flat Flange Lids, Suitable for reaction vessels with SCHOTT flat flange, with center neck socket NS 29/32, lateral necks with ground joint sockets or GL-thread. Bottom side with centring collar for non-slipping alignment on the vessel. All lateral c 1 * 1 items
Catalog # BOHLB210-22
Supplier:  Bohlender
Frequently Bought Together