ContactUsLinkComponent
Screw In Stopcocks, 3-Way, For connection to screw-in threads on BOLA Reactor Lids to interrupt the flow of connected tubes. Availab le as 2-way stopcock with straight bore and one GL-threaded connection or as 3-way stopcock with L-bore and two GL-th 1 * 1 items
Catalog # BOHLB156-08
Supplier:  Bohlender
undefined
Screw In Stopcocks, 3-Way, For connection to screw-in threads on BOLA Reactor Lids to interrupt the flow of connected tubes. Availab le as 2-way stopcock with straight bore and one GL-threaded connection or as 3-way stopcock with L-bore and two GL-th 1 * 1 items
Catalog # BOHLB156-08
Supplier:  Bohlender
Frequently Bought Together