ContactUsLinkComponent
Screw In Connector with Ground Joint, For connection to BOLA Reactor Lids. Connector with ground socket. With hexagonal gripping sur face in standard wrench size. 1 * 1 items
Catalog # BOHLB154-10
Supplier:  Bohlender
undefined
Screw In Connector with Ground Joint, For connection to BOLA Reactor Lids. Connector with ground socket. With hexagonal gripping sur face in standard wrench size. 1 * 1 items
Catalog # BOHLB154-10
Supplier:  Bohlender
Frequently Bought Together