0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Screw In Fittings GL, For connection to BOLA Reactor Lids as GL necks. With hexagonal gripping surface in standard wrench size. 1 * 1 items
Catalog # BOHLB152-14
Supplier:
Bohlender
Expand image
Screw In Fittings GL, For connection to BOLA Reactor Lids as GL necks. With hexagonal gripping surface in standard wrench size. 1 * 1 items
Catalog # BOHLB152-14
Supplier:
Bohlender
Frequently Bought Together
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
Screw In Fittings GL, For connection to BOLA Reactor Lids as GL necks. With hexagonal gripping surface in standard wrench size. 1 * 1 items
Supplier: Bohlender
für Schlauch Innen-Ø x Außen-Ø mm 1,6 x 3,2 1 * 1 items
Supplier: Bohlender