ContactUsLinkComponent
Reactor Lids, Suitable for glass reactors DN 150 with SCHOTT® flat flange. With screw in threads for connection of transition fitti ngs and a center neck NPT 1/2”. 1 * 1 items
Catalog # BOHLB150-22
Supplier:  Bohlender
undefined
Reactor Lids, Suitable for glass reactors DN 150 with SCHOTT® flat flange. With screw in threads for connection of transition fitti ngs and a center neck NPT 1/2”. 1 * 1 items
Catalog # BOHLB150-22
Supplier:  Bohlender
Frequently Bought Together