ContactUsLinkComponent
Double TemperatureProbes Lemo Compact EX, Two independent sensors PT 100 in a stainless steel tube (1.4571) encapsulated with stati c dissipative PTFE-EX. Collar ring Ø 12 mm, bore dia. 5 mm for earthing connection. The electric circuits are locally 1 * 1 items
Catalog # BOHLP1744-30
Supplier:  Bohlender
undefined
Double TemperatureProbes Lemo Compact EX, Two independent sensors PT 100 in a stainless steel tube (1.4571) encapsulated with stati c dissipative PTFE-EX. Collar ring Ø 12 mm, bore dia. 5 mm for earthing connection. The electric circuits are locally 1 * 1 items
Catalog # BOHLP1744-30
Supplier:  Bohlender
Frequently Bought Together