N-Methyl-N-phenyl-beta-alanine ethyl ester, 99% 1 * 25 g
Catalog # H61615.14
Supplier: Thermo Fisher Scientific
N-Methyl-N-phenyl-beta-alanine ethyl ester, 99% 1 * 25 g
Catalog # H61615.14
Supplier: Thermo Fisher Scientific
Frequently Bought Together
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific