ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 92 C, white backed, 40+70:0 ,1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 76mm, max.375x6-7mm, durable pigment, with Works Certificate with 4 test points 1 * 1 items
Catalog # AMARA301300-CB/WC
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 92 C, white backed, 40+70:0 ,1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 76mm, max.375x6-7mm, durable pigment, with Works Certificate with 4 test points 1 * 1 items
Catalog # AMARA301300-CB/WC
Supplier:  Amarell
Frequently Bought Together