ContactUsLinkComponent
High Select Top14 Abundant Protein Depletion Resin decreases the abundant protein and antibody components of human plasma samples, helping prepare the samples for mass spectrometry or 1D or 2D electrophoresis. The resin is provided here in a convenie 1 * 10 items
Catalog # PIERA36371
High Select Top14 Abundant Protein Depletion Resin decreases the abundant protein and antibody components of human plasma samples, helping prepare the samples for mass spectrometry or 1D or 2D electrophoresis. The resin is provided here in a convenie 1 * 10 items
Catalog # PIERA36371