ContactUsLinkComponent
CAPS, PHENOLIC SCREW, OPEN HOLE WITH SILICONE/PTFE LINERS FOR HEAVY WALL REACTION VIALS 20-400THREAD. BLACK 12/PACK. 1 * 12 items
Catalog # MICS98100-C
Supplier:  MICROSOLV
CAPS, PHENOLIC SCREW, OPEN HOLE WITH SILICONE/PTFE LINERS FOR HEAVY WALL REACTION VIALS 20-400THREAD. BLACK 12/PACK. 1 * 12 items
Catalog # MICS98100-C
Supplier:  MICROSOLV
Frequently Bought Together