ContactUsLinkComponent
8 components 100mg/l each of Al Au Cr Mo Na Sb Sn Te in HCl 2% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25
Catalog # CMPLRM002351L1
8 components 100mg/l each of Al Au Cr Mo Na Sb Sn Te in HCl 2% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25
Catalog # CMPLRM002351L1
Frequently Bought Together