ContactUsLinkComponent
21 components 100mg/l each of Ag As Ba Be Ca Cd Co Cu Fe K Li Mg Mn Ni P Pb Se Sr Tl V Zn in HNO3 2% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal lab
Catalog # CMPLRM002350L1
21 components 100mg/l each of Ag As Ba Be Ca Cd Co Cu Fe K Li Mg Mn Ni P Pb Se Sr Tl V Zn in HNO3 2% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal lab
Catalog # CMPLRM002350L1
Frequently Bought Together