ContactUsLinkComponent
Alcoholmeter richter & tralles, 0+35C, ca 340mm,0-100%, in 1/1 Vol%, tp 20C 1 * 1 items
Catalog # GERI0861
Supplier:  GERING
Alcoholmeter richter & tralles, 0+35C, ca 340mm,0-100%, in 1/1 Vol%, tp 20C 1 * 1 items
Catalog # GERI0861
Supplier:  GERING
Frequently Bought Together