ContactUsLinkComponent
EC HPLC column EC 75/3 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 75 mm, ID: 3 mm pack of 1 1 * 1 items
Catalog # MANA763433.30
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 75/3 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 75 mm, ID: 3 mm pack of 1 1 * 1 items
Catalog # MANA763433.30
Supplier:  MACHEREY-NAGEL
Frequently Bought Together