EC HPLC column EC 75/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 75 mm, ID: 2 mm pack of 1 1 * 1 items
MANA763433.20
EC HPLC column EC 75/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 75 mm, ID: 2 mm pack of 1 1 * 1 items
MANA763433.20
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection