ContactUsLinkComponent
EC HPLC column EC 50/4 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 50 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA763432.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 50/4 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 50 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA763432.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together