EC HPLC column EC 150/4.6 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 150 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA763436.46
Supplier: MACHEREY-NAGEL
EC HPLC column EC 150/4.6 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 150 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA763436.46
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Vial Racks
Supplier: VWR Collection
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
Exhaust filters, V3.0
Supplier: S.C.A.T.
Flat bottom chambers
Supplier: CAMAG
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Collecting trays, b.safe
Supplier: Bohlender
Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Cellulose microcrystalline for TLC plates
Supplier: Merck
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Screw caps for screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Funnel with ball and lid, ARNOLD, V2.0
Supplier: S.C.A.T.
CAP SCREW 24MM WHITE PP 24-400 PE FOAM / WHITE PTFE CLOSE TOP 100/PACK 1 * 100 items
Supplier: Thermo Fisher Scientific