ContactUsLinkComponent
C6010 Meter kit without electrodes: meter + 2x50 ml buffers (pH 4 and 7) + 50 ml electrolyte (3M KCl) + 50 ml conductivity standard (0.01 M KCl) + carrying case 1 * 1 items
Catalog # CONSC6010X
Supplier:  CONSORT
C6010 Meter kit without electrodes: meter + 2x50 ml buffers (pH 4 and 7) + 50 ml electrolyte (3M KCl) + 50 ml conductivity standard (0.01 M KCl) + carrying case 1 * 1 items
Catalog # CONSC6010X
Supplier:  CONSORT
Frequently Bought Together