ContactUsLinkComponent
Microslide superfrost cut pink glass cello ISO 8037/1 1,0mm 1 * 2.000 items
Catalog # MENZAA00008232E012
Supplier:  Epredia
Microslide superfrost cut pink glass cello ISO 8037/1 1,0mm 1 * 2.000 items
Catalog # MENZAA00008232E012
Supplier:  Epredia
Frequently Bought Together