ContactUsLinkComponent
Exhaust waste vapour condenser PELTRONIC for eight-cylinder CNT pumps (chemistry version), with assembly kit to connect to the pump, 100-120/200-230 V 50-60 Hz,without mains cable 1 * 1 items
Catalog # VACU22615511
Supplier:  VACUUBRAND
Exhaust waste vapour condenser PELTRONIC for eight-cylinder CNT pumps (chemistry version), with assembly kit to connect to the pump, 100-120/200-230 V 50-60 Hz,without mains cable 1 * 1 items
Catalog # VACU22615511
Supplier:  VACUUBRAND
Frequently Bought Together