DURAN® Safety Joints, Threaded Socket
Borosilicate glass 3.3, 160 mm, 45/40 NS, 50 mm, 3,5 mm
Catalog # SCOT286584503
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
NS joints with threaded socket, solid.
- Made from Borosilicate 3.3 glass
- Increased wall thickness
- Threaded sleeve prevents unintentional loosening of the ground joint
- Firmly seated ground joints can be loosened more easily here, the danger of glass breakage and associated risks is thus minimised
- Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range
- Tight closure due to standard ground joint
In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN® Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN® Safety Joints offer a system to separate jammed cones and sockets. This minimises the risk of breakage and related threats. The DURAN® cores with collar are designed for this purpose with an additionally formed glass ring.
Specifications
- Socket size:45/40 NS
- Tube ext.Ø:50 mm
- Wall thickness:3,5 mm
- Material:Borosilicate glass 3.3
- Seq:06
- Total length:160 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



