ContactUsLinkComponent
An ELISA kit for the detection of RAN (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 37.5pg/ml. 1 * 1 KIT
Catalog # GENOIT4963
Supplier:  G-Biosciences
An ELISA kit for the detection of RAN (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 37.5pg/ml. 1 * 1 KIT
Catalog # GENOIT4963
Supplier:  G-Biosciences
Frequently Bought Together