ContactUsLinkComponent
An ELISA kit for the detection of CAST (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Catalog # GENOIT5572
Supplier:  G-Biosciences
An ELISA kit for the detection of CAST (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Catalog # GENOIT5572
Supplier:  G-Biosciences
Frequently Bought Together