ContactUsLinkComponent
An ELISA kit for the detection of Total HSP-90 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.938ng/m l. 1 * 1 KIT
Catalog # GENOIT4670
Supplier:  G-Biosciences
An ELISA kit for the detection of Total HSP-90 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.938ng/m l. 1 * 1 KIT
Catalog # GENOIT4670
Supplier:  G-Biosciences
Frequently Bought Together